5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] solution staining reagent, for histology

Leverandør: Tonbo Biosciences

Synonymer: CFDA-SE, CFSE, Carboxyfluorescein diacetate succinimidyl ester, 5(6)-CFDA N-succinmidyl ester, 5(6)-Carboxyfluorescein diacetate succinimidyl ester, 5(6)-CFDA SE

13-0850-U500
13-0850-U500EA 330 NOK
13-0850-U500
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] solution staining reagent, for histology
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)]

CFSE is a non-fluorescent molecule that easily diffuses across cell membranes


Inside the cell, acetate groups are cleaved by intracellular esterases yielding a fluorescent molecule whose succinimidyl ester group covalently interacts with primary amines of intracellular proteins. CFSE is compatible with standard intracellular staining protocols using aldehyde fixation and saponin-based permeabilization. CFSE is widely used to track cell division and to monitor cell migration in vivo. CFSE can be detected using standard fluorescein filter sets by fluorescence microscopy or flow cytometry.


Store Undiluted at −20 °C; Protect from Light and Moisture

Formel: C₂₉H₁₉NO₁₁
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00037466
CAS-nummer: 150347-59-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR