Ghost Dye™
Leverandør: Tonbo Biosciences
13-0879-T100
13-0879-T100EA 615 NOK
13-0879-T100
Ghost Dye™
Ghost Dye™
Ghost Dye™ Violet 540 is an amine reactive viability dye that can be used to discriminate viable from non-viable mammalian cells in flow cytometry applications
  • Excitation Laser Violet (405 nm)
  • Emission (nm) 537
This dye irreversibly binds free amines available on the cell surface as well as intracellular free amines exposed in cells with compromised cell membranes. Necrotic cells with compromised membranes will react with significantly more Ghost Dye™ Violet 540 dye than viable cells in the same sample and therefore will exhibit much greater fluorescence intensity allowing exclusion of these cells from analysis.