Peqlab peqGOLD, DNA Isolation, Microspin Tissue DNA Mini Kits
Leverandør: VWR Chemicals
13-3096-02EA 6975 NOK
13-3096-02
Peqlab peqGOLD, DNA Isolation, Microspin Tissue DNA Mini Kits
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit
Fast, easy, and economical kit for isolation of genomic DNA from small sample volumes and sizes for consistent PCR and analytical test results.
  • No need for organic extractions
  • No need for time-consuming ethanol precipitations
  • Enzymatic degradation of cellular RNA
  • Efficient separation of inhibitors and contaminants
  • Fast, 20 min or less
Typical starting amount 10 mg tissue. Speed of mini column technology and specifically formulated buffers allow the purification of genomic DNA up to 60 kb.
Leveringsinformasjon: Kit contains peqGOLD Microspin LE DNA columns, 2 ml collection tubes, BL buffer, TL buffer, Proteinase K solution, HBC buffer, LPA buffer DNA wash buffer and elution buffer.