Peqlab peqGOLD, DNA Isolation, Blood And Tissue DNA Mini Kits

Leverandør: VWR Chemicals

13-3396-00EA 280 NOK
13-3396-00 13-3396-01 13-3396-02
Peqlab peqGOLD, DNA Isolation, Blood And Tissue DNA Mini Kits
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit

Fast, easy, and economical kit for isolation of DNA from a variety of samples including fresh or frozen cultured cells, tissue, blood, buccal swabs and saliva.


  • No need for organic extractions
  • No need for time-consuming alcohol precipitations
  • Enzymatic degradation of cellular RNA
  • Efficient separation of enzyme inhibitors and contaminants
  • High yields due to strong lysis
  • Fast, purification in less than 20 min (after lysis)


Typical starting amount should be 30 mg tissue, 5×10⁶ cultured cells, or up to 250 µl. Gives purified DNA suitable for almost any downstream application.


Leveringsinformasjon: Kit contains peqGOLD DNA Mini columns, 2 ml collection tubes, BL buffer, TL buffer, Proteinase K solution, HBC buffer, DNA wash buffer and elution buffer.

Bestill nå

SPESIFIKASJONER

Beskrivelse Blood and tissue DNA mini kit
Environmentally Preferable
Applikasjon
Target molecule DNA
Refine target molecule Total DNA
Sample type Bacteria,Blood,Biofluids (serum, plasma, saliva),Cultured cells,FFPE tissues,Human/animal cells and tissues
Method / Format Column (silica-based)
Prep size Mini
Prøvestørrelse 30 mg tissue, 5×10⁶ cultured cells, or up to 250 µl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR