Peqlab peqGOLD, RNA Isolation, Plant RNA Kit
Leverandør: VWR Chemicals
13-6627-01EA 3450 NOK
13-6627-01 13-6627-02
Peqlab peqGOLD, RNA Isolation, Plant RNA Kit
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit
Plant RNA Kit efficiently isolates RNA from a wide range of plant species and plant tissue, effectively removing genomic DNA, polysaccharides, phenol and other secondary metabolites. Starting amount can be up to 100 mg plant tissue. The isolated RNA is directly suitable for downstream applications.
  • Ideal for the simultaneous processing of multiple samples
  • DNA removing columns for efficient separation of DNA and cellular debris
  • No need for organic extractions
  • Immediate inactivation of endogenous and exogenous RNases
  • Favoured binding of RNA molecules >200 bases
  • Efficient separation of enzyme inhibitors and contaminants
  • Final elution in RNase-free water
Leveringsinformasjon: Kits contains peqGOLD RNA Mini columns, peqGOLS RNA Homogenizer columns, 2 ml collection tubes, NTL lysis buffer, SP buffer, RB buffer, RNA wash buffer 1 and nuclease-free water.