Peqlab peqGOLD, Plasmid Purification, Plasmid Miniprep Kit II
Leverandør: VWR Chemicals
13-6945-00EA 190 NOK
13-6945-00
Peqlab peqGOLD, Plasmid Purification, Plasmid Miniprep Kit II
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit
Isolate up to 40 to 70 µg of high quality plasmid DNA from up to 15 ml bacterial cultures in 30 minutes or less. Simplified with VWR® peqGold Mini Column II technology into three quick steps: Bind, wash and elute.
  • Safe, no phenol/chloroform extractions
  • Versatile with both spin and vacuum formats available
  • High quality DNA is suitable for a wide range of downstream applications
Leveringsinformasjon: Each kit contains peqGOLD DNA columns, 2 ml collection tubes, Solution I, Solution II, Solution III, HBC buffer, DNA wash buffer, RNase A and elution buffer.