Cobalt(II) fluoride, vannfri 98%

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: Cobalt difluoride, Cobaltous fluoride

13074.14 13074.22
13074.14EA 1125 NOK
13074.14 13074.22
Cobalt(II) fluoride, vannfri 98%
Cobalt(II) fluoride

Purity: 98% CAS No.: 10026-17-2 Molecular Formula: CoF2 Formula Weight: 96.93 Physical Form: Powder Density: 4.46 Melting Point: 1200° Boiling Point: 1400° Merck Reference: 14,2440 UN No.: 2923 MDL No.: MFCD00010941

Formel: CoF₂
MW: 96.93 g/mol
Kokepunkt: 1400 °C
Smeltepunkt: 1200
Tetthet: 4.46
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010941
CAS-nummer: 10026-17-2
EINECS: 233-061-9
FN: 2923
ADR: 8 III
Merck Indeks: 14,2440

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR