Fluorescein dinatriumsalt BAKER C.I. 45350, J.T.Baker®

Leverandør: Avantor

Synonymer: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium salt, Acid Yellow 73, Yellow No. 8, Fluorescein sodium salt, 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate

1314.0025EA 210 NOK
1314.0025
Fluorescein dinatriumsalt BAKER C.I. 45350, J.T.Baker®
Fluorescein dinatriumsalt
Formel: C₂₀H₁₀Na₂O₅
Smeltepunkt: > 300 °C
Tetthet: 1,601 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00167039
CAS-nummer: 518-47-8
EINECS: 208-253-0
Merck Indeks: 13,04185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance of solution passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR