Kalsiumklorid dihydrat, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor
Warning

1318.1000EA 2225 NOK
1318.1000 1318.0100
Kalsiumklorid dihydrat, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Kalsiumklorid dihydrat
Formel: CaCl₂·2H₂O
Kokepunkt: 1600 °C (1013 hPa)
Smeltepunkt: 176 °C
Tetthet: 1,85 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149613
CAS-nummer: 10035-04-8
EINECS: 233-140-8
Merck Indeks: 13,01660

Bestill nå

Spesifikasjon Testresultater

Assay min. 99.0 %
Ammonium (NH4) max. 0.005 %
Barium (Ba) max. 0.003 %
Potassium (K) max. 0.01 %
Magnesium (Mg) max. 0.05 %
Sodium (Na) max. 0.01 %
Strontium (Sr) max. 0.05 %
Insoluble in Water max. 0.005 %
Oxidizing Substances (as NO3) max. 0.003 %
pH of 5% Solution at 20°C 4.5 - 6.5
Sulfate (SO4) max. 0.005 %
Trace Impurities (in ppm)
Iron (Fe) max. 5
Heavy Metals (as Pb) max. 5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR