VWR® Cleanroom Stool
Leverandør: VWR Collection
132-1452EA 6275 NOK
132-1452
VWR® Cleanroom Stool
Stoler
This VWR® stool is suitable for clean rooms, certified and complete ESD safety. Featuring an easy and convenient height adjustment from any sitting position thanks to a ring release underneath the seat, which is protected from contamination by the soft cover.
  • 450 – 650 mm height adjustment
  • 400 mm seat diameter
  • Air cleanliness class 3 according to DIN EN ISO 14644-1
  • Exceeds the ESD requirements of DIN EN 61340-5-1
  • GS certificate for "tested safety"
The extra-large seat is made of synthetic black leather upholstery (60 mm high) and the rounded frontMore Product Information