Kalsiumklorid dihydrat, granular USP, Multi-Compendial, J.T.Baker®

Leverandør: Avantor
Warning

Synonymer: Calcium Chloride Dihydrate, Calcium dichloride

Management of Change (MOC) category = R

1335-07EA 11375 NOK
1335-07
Kalsiumklorid dihydrat, granular USP, Multi-Compendial, J.T.Baker®
Kalsiumklorid dihydrat
Formel: CaCl₂·2H₂O
MW: 147.01 g/mol
Kokepunkt: 1600
Smeltepunkt: 176
Tetthet: 1.85 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149613
CAS-nummer: 10035-04-8
EINECS: 233-140-8
Merck Indeks: 13,01660

Bestill nå

Spesifikasjon Testresultater

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
Meets J.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
USP - Identification Passes Test
USP - pH of 5% Solution at 25°C 4.5 - 9.2
USP - Iron, Aluminum, and Phosphate Passes Test
USP - Magnesium and Alkali Salts ≤ 1.0 %
USP - Assay (CaCl₂·2H₂O) 99.0 - 107.0 %
Endotoxin Concentration (EU/g) ≤ 10
EP/BP - Assay (CaCl₂·2H₂O) 97.0 - 103.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP/BP - Aluminum (Al) Passes Test
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Magnesium and Alkali Metals ≤ 0.5 %
JP - Assay (CaCl₂·2H₂O) 96.7 - 103.3 %
JP - Identification Passes Test
JP - pH (1 in 20) 4.5 - 9.2
JP - Heavy Metals (as Pb) ≤ 10 ppm
JP - Clarity and Color of Solution Passes Test
JP - Sulfate (SO₄) ≤ 0.024 %
JP - Hypochlorite Passes Test
JP - Iron, Aluminum or Phosphate Passes Test
JP - Barium (Ba) Passes Test
JP - Arsenic (As) ≤ 2 ppm
No Class 1,2,3 or other solvents are used or produced in the Passes Test
manufacturing or purification of the product. = 2 ppm
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR