Kalsiumklorid dihydrat 99.0-107.0%, CalPlus™ USP, Multi-Compendial, J.T.Baker®

Leverandør: Avantor
Warning


Management of Change (MOC) category = R

1336-01EA 1100 NOK
1336-01 1336-05 1336-07 1336-08 1336-09 1336-R
Kalsiumklorid dihydrat 99.0-107.0%, CalPlus™ USP, Multi-Compendial, J.T.Baker®
Kalsiumklorid dihydrat

Granular.


No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.

Formel: CaCl₂·2H₂O
Kokepunkt: 1600 °C (1013 hPa)
Smeltepunkt: 176 °C
Tetthet: 1,85 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149613
CAS-nummer: 10035-04-8
EINECS: 233-140-8
Merck Indeks: 13,01660

Bestill nå

Spesifikasjon Testresultater

Meets F.C.C. Requirements
Meets U.S.P Requirements
GMP Manufactured Product
Meets J.P. Chemical Specifications
Meets E.P. Chemical Specifications
Food GMP Manufactured Product
Meets B.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (CaCl₂·2H₂O) 99.0 - 107.0 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - pH of 5% Solution at 25°C 4.5 - 9.2
USP - Iron, Aluminum, and Phosphate Passes Test
USP - Magnesium and Alkali Salts ≤ 1.0 %
USP - Aluminum (Al)(µg/g) ≤ 1
Endotoxin Concentration (EU/g) ≤ 10
FCC - Assay (CaCl₂·2H₂O) 99.0 - 107.0 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Arsenic (As) ≤ 3 mg/kg
FCC - Magnesium and Alkali Salts ≤ 1.0 %
FCC - Lead (Pb) ≤ 5 mg/kg
FCC - Fluoride (F) ≤ 0.004 %
Appearance (White crystals or granules) Passes Test
EP/BP - Assay (CaCl₂·2H₂O) 97.0 - 103.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Sulfate (SO₄) ≤ 300 ppm
EP - Aluminum (Al) ≤ 1 ppm
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Magnesium and Alkali Metals ≤ 0.5 %
JP - Assay (CaCl₂·2H₂O) 96.7 - 103.3 %
JP - Identification Passes Test
JP - pH (1 in 20) 4.5 - 9.2
JP - Heavy Metals (as Pb) ≤ 10 ppm
JP - Clarity and Color of Solution Passes Test
JP - Sulfate (SO₄) ≤ 0.024 %
JP - Hypochlorite Passes Test
JP - Iron, Aluminum or Phosphate Passes Test
JP - Barium (Ba) Passes Test
JP - Arsenic (As) ≤ 2 ppm
No Class 1,2,3 or other solvents are used or produced in the ≤ 2 ppm
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.
Suitable for use in hemodialysis.

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR