Biological safety cabinets, class II, BioVanguard Green Line

Leverandør: CleanAir by Baker
NH501003 NH501001 NH601001 NH401003 NH401001 NH301001 NH601003
135-1717EA 0 NOK
135-1717 135-1715 135-1723 135-1709 135-1692 135-1705 135-1725
Biological safety cabinets, class II, BioVanguard Green Line
Avtrekksskap og kabinetter Sikkerhetskabinetter
BioVanguard Green Line is a Class II cabinet range which offers simplicity, robustness and high reliability and ensures the highest level of protection for the operator, product and environment, minimising the hazards inherent with working with agents assigned to biosafety levels one, two and three. It is designed for microbiological research with biological agents (e.g. bacteria, viruses, etc) and allergens.

  • Easy to install, easy to operate, easy to clean, easy to decontaminate and low maintenance
  • Microprocessor control with automatic compensation for
More Product Information
Bestill nå

SPESIFIKASJONER

Modell BioVanguard 5 BioVanguard 6 BioVanguard 4 BioVanguard 3
Environmentally Preferable
Filtre Pre-filter (EN 779): G3
Downflow HEPA filter (EN 1822): H14
Exhaust HEPA filter (EN 1822): H14
Pre-filter (EN 779): G3
Downflow HEPA filter (EN 1822): H14
Exhaust HEPA filter (EN 1822): H14
Pre-filter (EN 779): G3
Downflow HEPA filter (EN 1822): H14
Exhaust HEPA filter (EN 1822): H14
Pre-filter (EN 779): G3
Downflow HEPA filter (EN 1822): H14
Exhaust HEPA filter (EN 1822): H14
gjennomsnittlig lufthastighet 0,36 m/s 0,36 m/s 0,36 m/s 0,36 m/s
Belysningssystem 800 lux 1350 lux 1150 lux 950 lux
Støynivå 52 dB(A) 54 dB(A) 51 dB(A) 52 dB(A)
Effekt 0,199 kW 0,271 kW 0,156 kW 0,135 kW
Ext. W×D×H 1679×794×1602 mm 1984×794×1602 mm 1374×794×1602 mm 1072×794×1602 mm
Int. W×D×H 1485×550×744 mm 1790×550×744 mm 1180×550×744 mm 875×550×744 mm
Weight 275 kg 300 kg 215 kg 200 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR