Biological safety cabinets, class II, BioVanguard Green Line
Leverandør: CleanAir by Baker
NH501003 NH501001 NH601001 NH401003 NH401001 NH301001 NH601003
135-1717EA 0 NOK
135-1717 135-1715 135-1723 135-1709 135-1692 135-1705 135-1725
Biological safety cabinets, class II, BioVanguard Green Line
Avtrekksskap og kabinetter Sikkerhetskabinetter
BioVanguard Green Line is a Class II cabinet range which offers simplicity, robustness and high reliability and ensures the highest level of protection for the operator, product and environment, minimising the hazards inherent with working with agents assigned to biosafety levels one, two and three. It is designed for microbiological research with biological agents (e.g. bacteria, viruses, etc) and allergens.
  • Easy to install, easy to operate, easy to clean, easy to decontaminate and low maintenance
  • Microprocessor control with automatic compensation for