Gateway Accessory Kit for OPC™ UA Enablement of Bio Safety Cabinets
Leverandør: Thermo Scientific
50170006
135-2907EA 13463 NOK
135-2907
Gateway Accessory Kit for OPC™ UA Enablement of Bio Safety Cabinets
Avtrekksskap og kabinetter Sikkerhetskabinetter
Enable Open Platform Communication Unified Architecture (OPC™ UA) integration of Thermo Scientific™ Biological Safety Cabinets using this gateway accessory kit. Benefit from the full potential of incorporating laboratory equipment into your new or existing Distributed Control Systems (DCS), including paperless record keeping, remote monitoring, cloud analytics and more.
  • 21 CFR Part 11-compliance and GMP needs
  • Support audit requirements with enhanced data traceability
  • Real-time remote monitoring, workflow efficiencies, and assets and cost optimisation and