MCI GEL® CHP20P 75 - 150 μm, Supelco®
Leverandør: Merck
M-Clarity™ quality level = MQ100
13629-U 13630-U
13629-UEA 3325 NOK
13629-U 13630-U
MCI GEL® CHP20P 75 - 150 μm, Supelco®
MCI GEL® CHP20P
MCI GEL® CHP20P is a highly porous styrene-divinylbenzene polymer resin used as column packing material.
  • Surface area: ~500 m²/g
Polyaromatic adsorbent resin designed for biopharmaceutical separations: aromatic compounds, peptides, steroids, desalting, etc. Very good for reversed phase chromatography applications, even with nonaqueous solvents.