Cesium 1000 mg/L Cs in 5% salpetersyre (fra Cs₂CO₃) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

13825.AD 13825.AE 13825.AP
13825.ADEA 1075 NOK
13825.AD 13825.AE 13825.AP
Cesium 1000 mg/L Cs in 5% salpetersyre (fra Cs₂CO₃) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Cesium Standards

Molecular Formula: Cs₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134037


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR