Natrium 1000 mg/L Na in 5% salpetersyre (fra Na₂CO₃) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

13832.AD 13832.AE 13832.AP
13832.ADEA 835 NOK
13832.AD 13832.AE 13832.AP
Natrium 1000 mg/L Na in 5% salpetersyre (fra Na₂CO₃) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Sodium Standards

Molecular Formula: Na₂CO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085307


Sertifiseringer: Traceable to NIST

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00085307

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR