Hafnium 1000 mg/L Hf in 5% saltsyre (fra HfOCl₂) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

13843.ADEA 975 NOK
13843.AD 13843.AE
Hafnium 1000 mg/L Hf in 5% saltsyre (fra HfOCl₂) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Hafnium Standards

Molecular Formula: HfClO₂ in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011032


Sertifiseringer: Traceable to NIST

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00011032

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR