Indium 1000 mg/L In in 5% salpetersyre (fra In) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

13846.AD 13846.AE 13846.AP
13846.ADEA 1150 NOK
13846.AD 13846.AE 13846.AP
Indium 1000 mg/L In in 5% salpetersyre (fra In) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Indium Standards

Molecular Formula: In in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134048


Sertifiseringer: Traceable to NIST

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
MDL No. MFCD00134048

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR