Terbium 1000 mg/L Tb in 5% salpetersyre (fra Tb₄O₇) Specpure® enkeltgrunnstoffsstandard til ICP
Leverandør: Thermo Fisher Scientific
13860.AD 13860.AE 13860.AP
13860.ADEA 1100 NOK
13860.AD 13860.AE 13860.AP
Terbium 1000 mg/L Tb in 5% salpetersyre (fra Tb₄O₇) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Terbium Standards
Molecular formula: Tb₄O₇ in 5% HNO₃
Physical form: liquid
UN No.: 3264
MDL No.: MFCD00011256
Sertifiseringer: Traceable to NIST.