Neodymium 1000 mg/L Nd in 5% salpetersyre (fra Nd₂O₃) Specpure® enkeltgrunnstoffsstandard til ICP

Leverandør: Thermo Fisher Scientific

13882.AD 13882.AE 13882.AP
13882.ADEA 1150 NOK
13882.AD 13882.AE 13882.AP
Neodymium 1000 mg/L Nd in 5% salpetersyre (fra Nd₂O₃) Specpure® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Neodymium Standards

Molecular Formula: Nd₂O₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011130


Sertifiseringer: Traceable to NIST

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR