Rhodamin B, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Leverandør: Avantor
Danger

Synonymer: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride

1389.0100EA 680 NOK
1389.0100
Rhodamin B, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Rhodamin B
Formel: C₂₈H₃₁ClN₂O₃
Smeltepunkt: 199…201 °C
Tetthet: 1,31 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011931
CAS-nummer: 81-88-9
EINECS: 201-383-9
Merck Indeks: 13,08266

Bestill nå

Spesifikasjon Testresultater

Sensitivity as Indicator passes test
Solubility passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR