Transport trolleys, BC series
Leverandør: Metro International
BC2030-2DMB BC2030-3DG BC2030-3DMB BC2636-2DMB BC2636-3DMB
139-0055EA 6100 NOK
139-0055 139-0057 139-0058 139-0774 139-0775
Transport trolleys, BC series
Traller
Polymer utility carts consisting of at least two high density polyethylene, injection-moulded shelves and four chrome plated posts. Two shelf cart design allows user to add an intermediate shelf if deemed necessary. Cart will roll on four (102 mm diameter thermoplastic), mark resistant swivel/stem castors.
  • Smooth surfaces are designed to resist staining and wipe clean easily
  • Corrosion-proof shelves
  • Sturdy construction: Each shelf can hold up to 68 kg; each unit can hold up to 181 kg
  • Easy to adjust: Optional centre shelf adjusts on 25 mm increments