Lanyard, Miller® Titan™ A3
Leverandør: HONEYWELL SAFETY
1013430
139-0237EA 1375 NOK
139-0237
Lanyard, Miller® Titan™ A3
Fallvern Falltau
Webbing lanyard with shock absorber that reduces the impact force to less than 6 kN in the event of a fall.
  • Polyester webbing (30 mm wide)
  • With screwgate karabiner and scaffold hook (63 mm opening)
Sertifiseringer: EN 362