Lanyard, Miller® Titan™ Y
Leverandør: HONEYWELL SAFETY
1013431
139-0238EA 1875 NOK
139-0238
Lanyard, Miller® Titan™ Y
Fallvern Falltau
Y-lanyard with two scaffold hooks for alternate anchoring at one end and a zinc plated karabiner at the other end. Featuring an integrated shock absorber that reduces the impact force to less than 6 kN in the event of a fall.
  • Polyester webbing and polyamide shock absorber
  • With screwgate karabiner and two scaffold hooks (63 mm opening)
Sertifiseringer: EN 362