Trolleys, basket, KR 60-60
Leverandør: HUPFER METALLWERKE
7500196
139-0800EA 13150 NOK
139-0800
Trolleys, basket, KR 60-60
Traller
These trolleys made of stainless steel can take baskets, containers and bins made from polypropylene.
  • Stable, mobile round tube construction
  • Mobile on four swivel castors, two of them with brake
  • Galvanised-chromed housings with electro-conductive double ball-bearing
  • Two bumpers protect trolley against damage