Hupfer Variocart® MVC 6×4/3 KST/gr Transport Trolley
Leverandør: HUPFER METALLWERKE
MVC6X4/3KST/GR
139-0816EA 9775 NOK
139-0816
Hupfer Variocart® MVC 6×4/3 KST/gr Transport Trolley
Traller
Demountable transport trolley made of high-quality stainless steel with deep shelves.
  • Height grid: 66,5 mm
  • On-board loading: 100 kg
Demountable transport trolley in a sturdy, self-supporting and hygienic design made of high-quality stainless steel round tube frame Ø25 mm with adjustable, height-adjustable shelves, bevelled on all sides, with a sturdy clamping device. Four polyethylene disc bumpers serve as collision protection. Runs on four corrosion-resistant swivel castors Ø125 mm, two with locks, grey polymer housing with air-identic polymer wheelMore Product Information