Steps, mobile
Leverandør: WEDO WERNER DORSCH
2151
139-9806EA 1800 NOK
139-9806
Steps, mobile
Stiger og krakker
Robust aluminium 2×2 steps. Fitted with castors to permit movement in every direction. All edges are protected by plastic caps to prevent injuries.
  • Castors with spring bearing
  • Robust and non-slip
  • The stepladder sinks down into an immobile position as soon as weight is put on the ribbed treads, thus allowing safe use
Maximum load: 150 kg
Sertifiseringer: TÜV and GS tested in compliance with DIN 4569