Sodium stannate hydrate, Reagent Grade

Leverandør: Thermo Fisher Scientific
Warning

14021.22 14021.36
14021.22EA 640 NOK
14021.22 14021.36
Sodium stannate hydrate, Reagent Grade
Sodium stannate hydrate

Sodium tin(IV) oxide is used as a mordant, fire retardant textiles.


CAS No.: 12058-66-1 Molecular Formula: Na2SnO3.xH2O Formula Weight: 212.67(anhy) Physical Form: Crystalline Sensitivity: Air Sensitive Odour: Odourless Merck Reference: 14,8677 MDL No.: MFCD00014241

Formel: Na₂O₃Sn·H₂O
Smeltepunkt: 140 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00014241
CAS-nummer: 12058-66-1
EINECS: 235-030-5
Merck Indeks: 14,8677

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR