Glass Microfibre Cloth, Hygen™
Leverandør: Rubbermaid Commercial Products
FGQ63000BL00
141-0122EA 980 NOK
141-0122
Glass Microfibre Cloth, Hygen™
Papirservietter Rengjøringskluter for gjenbruk Kluter til generell bruk
The Rubbermaid Commercial HYGEN™ Mirror and Glass Cloths feature premium Microfibre construction for scratch-free and lint-free cleaning.
  • Premium Microfibre construction with quality weave and double-finished edges
  • Designed for scratch- and lint-free cleaning of glass and mirrored surfaces
  • Durable for up to 500 launderings (up to 200 with chlorine)
  • Chlorine safe in hot and cold water