Foam Pistol Bottle, Nito Clean
Leverandør: WERNER MERTZ
0705727
141-0285EA 2275 NOK
141-0285
Foam Pistol Bottle, Nito Clean
Flasker Sprayflasker
Foam gun for applying a stable foam carpet on horizontal and vertical surfaces. The dosage can be adjusted.
For use with cleaning agents. Can be connected to a standard water tap.