Vaskemiddel for manuell rengjøring av glassvarer, pulver, neodisher® PM 5
Leverandør: Dr. Weigert
440281
141-0941EA 7575 NOK
141-0941
Vaskemiddel for manuell rengjøring av glassvarer, pulver, neodisher® PM 5
Rengjøringsløsninger Detergenter og overflatevasking
For manual cleaning of laboratory glassware and surgical instruments in hospitals, medical and biological laboratories, and food industry laboratories.
  • Mildly alkaline powder for washing surgical instruments and laboratory glassware by hand
  • Components: <5% non-ionic surfactants, anionic surfactants, bleaching agents (percarbonates); >30% phosphates
  • Dosage: 0,5 to 2%