Cleanroom vacuum cleaner, EMI-CWR
Leverandør: TIGER-VAC
110110B
141-1066EA 33600 NOK
141-1066
Cleanroom vacuum cleaner, EMI-CWR
Vakuumrensere
Designed for general clean up in contamination controlled environments, cleanroom compatible and ideal for pharmaceutical industry.
  • Four stage filtration system includes a ULPA filter (U15); double cloth filter, dome filter and disposable filter bag
  • An internal HEPA filter H14 is available as an optional 5th stage of filtration
  • Single phase through-flow motor
  • Dual speed control to regulate suction
  • Based on a four wheel dolly for easy manouvreability
  • It also incorporates a thermo-protector which prevents the motor from overheating if there is a blockage in