Vacuum cleaner, MRV-8 POLY
Leverandør: TIGER-VAC
110116B
141-1067EA 34875 NOK
141-1067
Vacuum cleaner, MRV-8 POLY
Vakuumrensere
This unit is designed for mercury recovery and includes Mersorb® brand activated carbon for the adsorption of mercury vapours.
Construction: Polyethylene recovery tank
  • Through-flow motor
  • EMI/RFI shielded
  • Timer (hour counter) included
  • Available with an optional variable speed controller
  • Based on a four wheel dolly Optional suction cups available
This unit is equipped with a true ULPA filter (U15) located upstream of the motor. It is EMI/RFI shielded, is equipped with an electronic filter suppresser which reduces electromagnetic interference (EMI), andMore Product Information