Industrial vacuum cleaner, EMI 1000(CV)
Leverandør: TIGER-VAC
110132B
141-1068EA 37575 NOK
141-1068
Industrial vacuum cleaner, EMI 1000(CV)
Vakuumrensere
Industrial vacuum cleaner EMI-1000 (CV) dry recovery with safe containment canister stainless steel type 316 2B powerhead, filter chamber and wheel base.
  • Designed for the safe collection, containment and disposal of potent compounds
  • Single phase through-flow motor
  • Electropolished version is available
  • Dual speed control to regulate suction
  • Based on a 4-wheel dolly for easy manoeuvrability
Ideal for the pharmaceutical industry, this unit is equipped with a true ULPA filter (U15) located downstream of the motor and a HEPA filter H14 located upstream. TheMore Product Information