Cleanroom vacuum cleaner, CR-4D
Leverandør: TIGER-VAC
110136B
141-1069EA 27400 NOK
141-1069
Cleanroom vacuum cleaner, CR-4D
Vakuumrensere
Designed for general clean up in contamination controlled environments, cleanroom compatible and ideal for pharmaceutical industry.
  • Four stage filtration system includes a ULPA Filter U15; double cloth filter, dome filter and disposable filter bag
  • An internal HEPA filter HEPA H14 is available as an optional 5th stage of filtration
  • Single phase thru-flow motor
  • Recovery capacity: 15,2 litres in recovery tank and 9,5 litres in disposible filter bag
  • Based on a four wheel dolly for easy manoeuvrability
This unit is equipped with a true ULPA filter (U15)More Product Information