Cleanroom vacuum cleaner, MV-1CR (HH)
Leverandør: TIGER-VAC
110735B
141-1071EA 22350 NOK
141-1071
Cleanroom vacuum cleaner, MV-1CR (HH)
Vakuumrensere
Designed for general clean up in contamination controlled environments, cleanroom compatible and ideal for pharmaceutical industry.
  • 4-stage filtration system includes a ULPA filter (U15); double cloth filter, dome filter and disposable filter bag
  • Special filter lined with stainless steel is available for the revovery broken wafers and sharp object
  • Single phase through-flow motor
  • Recovery capacity: 2 litres in disposible filter bag
This unit is equipped with a true ULPA filter (U15) located upstream of the motor. It is EMI/RFI shielded, equipped withMore Product Information