Cleanroom vacuum cleaner, MV-1CR (BP)
Leverandør: TIGER-VAC
110737B
141-1072EA 29250 NOK
141-1072
Cleanroom vacuum cleaner, MV-1CR (BP)
Vakuumrensere
Designed for a variety of cleanroom environments and mini environments, including semiconductor/microelectronic, pharmaceutical. Cleanroom compatible.
  • 4-stage filtration system includes a U15 filter, cloth filter, dome filter and disposable filter bag
  • Special filter lined with stainless steel is available for the recovery of broken wafers and sharp objects
  • Single phase through-flow motor
  • Recovery capacity of 5,7 litres in disposable filter bag
  • Equipped with a backpack harness
This unit is equipped with a true ULPA filter (U15) located upstream of theMore Product Information