Cleanroom vacuum cleaner, CWR-10 (PHARMA)
Leverandør: TIGER-VAC
110535B240V
141-1212EA 0 NOK
141-1212
Cleanroom vacuum cleaner, CWR-10 (PHARMA)
Vakuumrensere
This wet and dry recovery vacuum cleaner is cleanroom compatible and can be used in pharmaceutical industry. It has been designed for the recovery of broken ampoules and glass in contamination controlled environments. The broken ampoules and glass are separated from liquid and they are recovered in an autoclavable sieve basket made of electropolished stainless steel AISI 304.
  • For dry and wet recovery
  • Single phase by-pass motor
  • Based on a 4-wheel dolly for easy maneuverability
  • Autoclavable sieve basket with stainless steel mesh filter
  • Recovery capacity: 28,5