Cleanroom mops, MicroMop
Leverandør: PFENNIG REINIGUNGSTECHNIK
2700202
141-1392EA 170 NOK
141-1392
Cleanroom mops, MicroMop
Mopper
Mop for all smooth hard floorings. Made up of 100% PES super microfiber.
  • For wet and damp cleaning in the single-step wiping process
  • Also suitable for dry-cleaning, sealed parquet and laminate flooring
  • Backing with cover sheet and belt bag extremely tear