Compact bucket systems, PP
Leverandør: CONTEC
2940U2B UBKT0008 2940P2B PB6BS-3 2940U3B
141-1428EA 40625 NOK
141-1428 141-1436 141-1427 141-1435 141-1430
Compact bucket systems, PP
Traller
The autoclavable "Compact Bucket System" is an all-in-one solution to make cleaning and disinfecting easier and more efficient. The trolley is designed for use with Contec's VertiKlean® MAX™, VertiKlean® MAX™ Sealed Edge, EasyCurve™ and QuickTask™ Mopping Systems.
  • Compact polypropylene buckets with integral drip cap keep buckets in place to prevent spilling
  • Mop removal tool for VertiKlean® MAX™ and VertiKlean® MAX™ Sealed Edge mops
  • Graduated PP buckets available in three colours to denote waste, rinse, and clean solution
  • Utility handle basket provides