Autoclavable systembox EasyMop GMP with MopJump-System
Leverandør: PFENNIG REINIGUNGSTECHNIK
3500200 3500210 3500051 3500200QR
141-0952EA 8525 NOK
141-0952 141-0953 141-0951 141-1469
Autoclavable systembox EasyMop GMP with MopJump-System
Mopper
Autoclavable systembox EasyMop GMP and MopJump-System:
The newly developed 'EasyMop GMP' system box ensures compliance with regulations governing the cleaning and desinfecting of cleanrooms, and offers enormous advantages regarding its application.
  • All assembly components for use in pharmaceutical cleanrooms are certified
  • All measurements are precisely adapted to the intended use
  • The 'MopJump' system with displacement area allows easy impregnation of the mop head as well as a contact-free pick up
  • Triple-sealing ensures that the system box is securely