Industrial vacuum cleaner, backpack, T1-3L BP HEPA
Leverandør: TIGER-VAC
112212B
141-1549EA 8275 NOK
141-1549
Industrial vacuum cleaner, backpack, T1-3L BP HEPA
Vakuumrensere
This vacuum cleaner is equipped with a backpack harness, it is lightweight and ergonomic. It is designed for general cleanup. The construction is all plastic.
  • 3-stage filtration system: HEPA filter H14, cloth filter and disposable filter bag
  • Single phase thru-flow heavy duty motor
  • Ligthweight and portable
  • Recovery capacity: 3 L in disposable filter bag
  • Ready to use: filter and tool kit included
This unit is equipped with a true HEPA filter (H14) located upstream of the motor.
Note: For dry recovery only.
Sertifiseringer: HEPA filter tested: IESTMore Product Information