VWR® USC, Ultrasonic Cleaning Baths
Leverandør: VWR Collection
142-0087EA 16225 NOK
142-0087 142-0094
VWR® USC, Ultrasonic Cleaning Baths
Ultralydbad
Cleaning via ultrasound has been one of the most effective methods of freeing parts with complex geometry, such as fins, under cuts, bore holes or pocket holes from residues (contaminants).
  • High performance PZT ultrasonic converter with ceramics technology
  • Homogeneous distribution of ultrasound in the cleaning trough
  • Lower noise development through higher frequency
  • Robust heating with integrated run-dry protection
  • Thermostat adjustable up to 80 °C
  • Stainless steel tank material and cladding
The physical property of imploding vacuum bubbles causesMore Product Information