Alkaline cleaner, TICKOPUR R 33
Leverandør: BANDELIN-ELECTRONIC
883
142-1913EA 565 NOK
142-1913
Alkaline cleaner, TICKOPUR R 33
Rengjøringsløsninger Detergenter og overflatevasking
Universal cleaner is a corrosion protectant for service, industry, technology and laboratory with gentle cleaning. It can be used to clean glass, metal, ceramics, plastic, rubber and removes grease, oils, wax, silicone films, protein, ink residue, etc.
  • Mildly alkaline
  • Dosage 1 to 5%