Cleaning agents for use in ultrasonic baths, TICKOPUR TR3, Bandelin/basan
Leverandør: BANDELIN-ELECTRONIC
923 937
142-2015EA 550 NOK
142-2015 BAND937
Cleaning agents for use in ultrasonic baths, TICKOPUR TR3, Bandelin/basan
Rengjøringsløsninger Detergenter og overflatevasking
The cavitation improving and material protecting features allow a maximum effect of the ultrasonic cleaning.
  • Shortens the cleaning time significantly
  • Special cleaner based on citric acid, mildly acidic
  • Free from phosphate and have corrosion protection
Used for cleaning, rust removal, decalcification and degreasing of assembled and disassembled machines, tools and devices made of steel, stainless steel, non-ferrous, precious and light metals, glass, ceramics, plastics and rubber. Contaminants such as mineral residues, drifting rust, grease, oils, waxes,More Product Information