Antimony standard dissolved in oil, 1.000 µg/g Sb in mineral oil Specpure® til wear metal analysis
Leverandør: Thermo Fisher Scientific
14207.18
14207.18EA 1100 NOK
14207.18
Antimony standard dissolved in oil, 1.000 µg/g Sb in mineral oil Specpure® til wear metal analysis
Standarder Kationstandarder Antimony Standards
Physical form: liquid
MDL No.: MFCD00134030