Vann ACS for ultratrace analysis, Supelco®

Leverandør: Merck

Synonymer: Hydrogen oxide

M-Clarity™ quality level = MQ100

14211-1L-F
14211-1L-FEA 1880 NOK
14211-1L-F
Vann ACS for ultratrace analysis, Supelco®
Vann
Formel: H₂O
Kokepunkt: 100 °C (1013 hPa)
Smeltepunkt: 0 °C
Tetthet: 1 g/cm³ (4 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011332
CAS-nummer: 7732-18-5
EINECS: 231-791-2
Merck Indeks: 13,10098
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb883be5463dc4ac0766c79568501083468ed2c4&vid=69872816d497aabc2fff7251882830e91199263b&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR