Sodium standard dissolved in oil, 1.000 µg/g Na in mineral oil Specpure® til wear metal analysis

Leverandør: Thermo Fisher Scientific

14212.18
14212.18EA 1225 NOK
14212.18
Sodium standard dissolved in oil, 1.000 µg/g Na in mineral oil Specpure® til wear metal analysis
Standarder Kationstandarder Sodium Standards

Physical Form: Liquid
MDL No.: MFCD00085307

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR